N,N-Diethyl-3-pyrrolidinamine dihydrochloride
Supplier: Matrix Scientific
051523-500MG
051523-2.5G
101895-986EA
838.46
USD
101895-986
101895-984
N,N-Diethyl-3-pyrrolidinamine dihydrochloride
N,N-Diethyl-3-pyrrolidinamine dihydrochloride
Hazard: Irritant/Stored under Argon
Formula:
C₈H₂₀Cl₂N₂ |
MDL Number:
MFCD13561580 CAS Number: 20984-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...