3-(1-Pyrrolidinyl)pyrrolidine dihydrochloride
Supplier: Matrix Scientific
051522-2.5G
051522-500MG
101895-982EA
838.46
USD
101895-982
101895-980
3-(1-Pyrrolidinyl)pyrrolidine dihydrochloride
3-(1-Pyrrolidinyl)pyrrolidine dihydrochloride
Hazard: Irritant
Formula:
C₈H₁₈Cl₂N₂ |
MDL Number:
MFCD13561579 CAS Number: 267241-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...