2-(4-Piperidinyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Supplier: Matrix Scientific

051516-500MG 051516-2.5G
101895-956EA 364.62 USD
101895-956 101895-958
2-(4-Piperidinyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
2-(4-Piperidinyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂
MDL Number: MFCD06796139
CAS Number: 871113-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR