N-Cyclohexyl-N-ethyl-3-piperidinamine dihydrochloride
Supplier: Matrix Scientific
051487-500MG
051487-2.5G
101895-842EA
838.46
USD
101895-842
101895-840
N-Cyclohexyl-N-ethyl-3-piperidinamine dihydrochloride
N-Cyclohexyl-N-ethyl-3-piperidinamine dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₈Cl₂N₂ |
MDL Number:
MFCD13561556 CAS Number: 1220034-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...