2-[4-(4-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051461-2.5G 051461-500MG
101895-736EA 364.62 USD
101895-736 101895-738
2-[4-(4-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride
2-[4-(4-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₇Cl₂N₃O
MDL Number: MFCD13561530
CAS Number: 1220034-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR