2-[Ethyl(4-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051446-500MG 051446-2.5G
101895-678EA 838.46 USD
101895-678 101895-676
2-[Ethyl(4-piperidinylmethyl)amino]-1-ethanol dihydrochloride
2-[Ethyl(4-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂O
MDL Number: MFCD13561516
CAS Number: 1219957-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR