N-Benzyl-N-(3-piperidinylmethyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051436-500MG 051436-2.5G
101895-636EA 364.62 USD
101895-636 101895-638
N-Benzyl-N-(3-piperidinylmethyl)-1-ethanamine dihydrochloride
N-Benzyl-N-(3-piperidinylmethyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₆Cl₂N₂
MDL Number: MFCD13561508
CAS Number: 1220030-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR