1-(3-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
Supplier: Matrix Scientific
051435-2.5G
051435-500MG
101895-634EA
838.46
USD
101895-634
101895-632
1-(3-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
1-(3-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄Cl₂N₂ |
MDL Number:
MFCD13561507 CAS Number: 1220021-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...