N-1-Benzyl-4-bromo-N-1-methyl-1,2-benzenediamine
Supplier: Matrix Scientific
051401-2.5G
051401-500MG
101895-498EA
838.46
USD
101895-498
101895-496
N-1-Benzyl-4-bromo-N-1-methyl-1,2-benzenediamine
N-1-Benzyl-4-bromo-N-1-methyl-1,2-benzenediamine
Hazard: Irritant
Formula:
C₁₄H₁₅BrN₂ |
MDL Number:
MFCD09941367 CAS Number: 1017046-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...