1-(2-Amino-4-bromophenyl)-4-piperidinol
Supplier: Matrix Scientific
051397-2.5G
051397-500MG
101895-482EA
838.46
USD
101895-482
101895-480
1-(2-Amino-4-bromophenyl)-4-piperidinol
1-(2-Amino-4-bromophenyl)-4-piperidinol
Hazard: Irritant
Formula:
C₁₁H₁₅BrN₂O |
MDL Number:
MFCD09933681 CAS Number: 1016777-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...