5-Bromo-2-(3-methyl-1-piperidinyl)aniline
Supplier: Matrix Scientific
051391-2.5G
051391-500MG
101895-458EA
838.46
USD
101895-458
101895-456
5-Bromo-2-(3-methyl-1-piperidinyl)aniline
5-Bromo-2-(3-methyl-1-piperidinyl)aniline
Hazard: Irritant
Formula:
C₁₂H₁₇BrN₂ |
MDL Number:
MFCD09942167 CAS Number: 1016806-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...