N-4-Benzyl-N-4-ethyl-2-methyl-1,4-benzenediamine

Supplier: Matrix Scientific

051378-500MG 051378-2.5G
101895-406EA 838.46 USD
101895-406 101895-404
N-4-Benzyl-N-4-ethyl-2-methyl-1,4-benzenediamine
N-4-Benzyl-N-4-ethyl-2-methyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₆H₂₀N₂
MDL Number: MFCD12410394
CAS Number: 757166-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR