4-(2,3-Dihydro-1H-indol-1-yl)-2-methylphenylamine

Supplier: Matrix Scientific

051376-500MG 051376-2.5G
101895-398EA 1934.49 USD
101895-398 101895-396
4-(2,3-Dihydro-1H-indol-1-yl)-2-methylphenylamine
4-(2,3-Dihydro-1H-indol-1-yl)-2-methylphenylamine

Hazard: Irritant

Formula: C₁₅H₁₆N₂
MDL Number: MFCD12410433
CAS Number: 1154319-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR