2-Methyl-N-4,N-4-dipropyl-1,4-benzenediamine

Supplier: Matrix Scientific

051369-2.5G 051369-500MG
101895-368EA 272.55 USD
101895-368 101895-370
2-Methyl-N-4,N-4-dipropyl-1,4-benzenediamine
2-Methyl-N-4,N-4-dipropyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₃H₂₂N₂
MDL Number: MFCD12169088
CAS Number: 105294-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR