1-(4-Amino-3-methylphenyl)-4-piperidinol
Supplier: Matrix Scientific
051368-500MG
051368-2.5G
101895-366EA
838.46
USD
101895-366
101895-364
1-(4-Amino-3-methylphenyl)-4-piperidinol
1-(4-Amino-3-methylphenyl)-4-piperidinol
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O |
MDL Number:
MFCD12169146 CAS Number: 161282-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...