2-Methyl-4-(4-methyl-1-piperazinyl)aniline
Supplier: Matrix Scientific
051364-500MG
051364-2.5G
101895-348EA
272.55
USD
101895-348
101895-350
2-Methyl-4-(4-methyl-1-piperazinyl)aniline
2-Methyl-4-(4-methyl-1-piperazinyl)aniline
Hazard: Irritant
Formula:
C₁₂H₁₉N₃ MW: 205.31 g/mol |
MDL Number:
MFCD06335528 CAS Number: 16154-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...