2-[4-Amino(ethyl)-3-methylanilino]-1-ethanol

Supplier: Matrix Scientific

051359-500MG 051359-2.5G
101895-328EA 364.62 USD
101895-328 101895-330
2-[4-Amino(ethyl)-3-methylanilino]-1-ethanol
2-[4-Amino(ethyl)-3-methylanilino]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₈N₂O
MDL Number: MFCD00476518
CAS Number: 2359-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR