2-Methyl-4-(1-piperidinyl)aniline dihydrochloride

Supplier: Matrix Scientific

051355-500MG 051355-2.5G
101895-314EA 950.81 USD
101895-314 101895-312
2-Methyl-4-(1-piperidinyl)aniline dihydrochloride
2-Methyl-4-(1-piperidinyl)aniline dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₀Cl₂N₂
MDL Number: MFCD24849141
CAS Number: 1449117-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR