2-(4-Amino-3-dimethylanilino)-1-ethanol

Supplier: Matrix Scientific

051354-2.5G 051354-500MG
101895-310EA 838.46 USD
101895-310 101895-308
2-(4-Amino-3-dimethylanilino)-1-ethanol
2-(4-Amino-3-dimethylanilino)-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₆N₂O
MDL Number: MFCD12169213
CAS Number: 63123-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR