N-1-Benzyl-N-1-ethyl-2-fluoro-1,4-benzenediamine

Supplier: Matrix Scientific

051349-2.5G 051349-500MG
101895-290EA 942.58 USD
101895-290 101895-288
N-1-Benzyl-N-1-ethyl-2-fluoro-1,4-benzenediamine
N-1-Benzyl-N-1-ethyl-2-fluoro-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₅H₁₇FN₂
MDL Number: MFCD11195371
CAS Number: 1038733-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR