4-(2,3-Dihydro-1H-indol-1-yl)-3-fluorophenylamine

Supplier: Matrix Scientific

051347-500MG 051347-2.5G
101895-280EA 166.15 USD
101895-280 101895-282
4-(2,3-Dihydro-1H-indol-1-yl)-3-fluorophenylamine
4-(2,3-Dihydro-1H-indol-1-yl)-3-fluorophenylamine

Hazard: Irritant

Formula: C₁₄H₁₃FN₂
MDL Number: MFCD08699584
CAS Number: 937597-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR