N-1-Benzyl-2-fluoro-N-1-methyl-1,4-benzenediamine
Supplier: Matrix Scientific
051343-2.5G
051343-500MG
101895-266EA
1161.79
USD
101895-266
101895-264
N-1-Benzyl-2-fluoro-N-1-methyl-1,4-benzenediamine
N-1-Benzyl-2-fluoro-N-1-methyl-1,4-benzenediamine
Hazard: Irritant
Formula:
C₁₄H₁₅FN₂ |
MDL Number:
MFCD11193230 CAS Number: 832099-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...