4-(2-Ethyl-1-piperidinyl)-3-fluoroaniline

Supplier: Matrix Scientific

051342-500MG
101895-260EA 272.55 USD
101895-260
4-(2-Ethyl-1-piperidinyl)-3-fluoroaniline
4-(2-Ethyl-1-piperidinyl)-3-fluoroaniline

Hazard: Irritant

Formula: C₁₃H₁₉FN₂
MDL Number: MFCD11624795
CAS Number: 1094863-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR