2-Fluoro-N-1,N-1-dipropyl-1,4-benzenediamine

Supplier: Matrix Scientific

051340-500MG 051340-2.5G
101895-254EA 838.46 USD
101895-254 101895-252
2-Fluoro-N-1,N-1-dipropyl-1,4-benzenediamine
2-Fluoro-N-1,N-1-dipropyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₂H₁₉FN₂
MDL Number: MFCD10691038
CAS Number: 1092276-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR