4-(1-Azepanyl)-3-fluoroaniline
Supplier: Matrix Scientific
051337-500MG
051337-2.5G
101895-242EA
838.46
USD
101895-242
101895-240
4-(1-Azepanyl)-3-fluoroaniline
4-(1-Azepanyl)-3-fluoroaniline
Hazard: Irritant
Formula:
C₁₂H₁₇FN₂ |
MDL Number:
MFCD08699583 CAS Number: 250371-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...