4-(4-Benzyl-1-piperazinyl)-3-fluorophenylamine

Supplier: Matrix Scientific

051336-500MG 051336-2.5G
101895-238EA 838.46 USD
101895-238 101895-236
4-(4-Benzyl-1-piperazinyl)-3-fluorophenylamine
4-(4-Benzyl-1-piperazinyl)-3-fluorophenylamine

Hazard: Irritant

Formula: C₁₇H₂₀FN₃
MDL Number: MFCD08699565
CAS Number: 937597-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR