3-Fluoro-4-(1-pyrrolidinyl)aniline

Supplier: Matrix Scientific

051325-500MG 051325-2.5G
101895-192EA 364.62 USD
101895-192 101895-194
3-Fluoro-4-(1-pyrrolidinyl)aniline
3-Fluoro-4-(1-pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₃FN₂
MW: 180.23 g/mol
MDL Number: MFCD07403468
CAS Number: 93246-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR