4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-methylaniline

Supplier: Matrix Scientific

051319-2.5G 051319-500MG
101895-170EA 838.46 USD
101895-170 101895-168
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-methylaniline
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₈N₂
MDL Number: MFCD11214010
CAS Number: 1094746-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR