(N-1,N-1)-2-Trimethyl-1,4-benzenediamine

Supplier: Matrix Scientific

051295-2.5G 051295-500MG
101895-074EA 437.05 USD
101895-074 101895-072
(N-1,N-1)-2-Trimethyl-1,4-benzenediamine
(N-1,N-1)-2-Trimethyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₉H₁₄N₂
MDL Number: MFCD09965938
CAS Number: 27746-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR