4-[3,4-Dihydro-1(2H)-quinolinyl]-3-(trifluoromethyl)aniline
Supplier: Matrix Scientific
051292-500MG
101895-060EA
364.62
USD
101895-060
4-[3,4-Dihydro-1(2H)-quinolinyl]-3-(trifluoromethyl)aniline
4-[3,4-Dihydro-1(2H)-quinolinyl]-3-(trifluoromethyl)aniline
Hazard: Irritant
Formula:
C₁₆H₁₅F₃N₂ |
MDL Number:
MFCD13561469 CAS Number: 1184441-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...