1-[4-Amino-2-(trifluoromethyl)phenyl]-4-piperidinol

Supplier: Matrix Scientific

051283-500MG 051283-2.5G
101895-026EA 838.46 USD
101895-026 101895-024
1-[4-Amino-2-(trifluoromethyl)phenyl]-4-piperidinol
1-[4-Amino-2-(trifluoromethyl)phenyl]-4-piperidinol

Hazard: Irritant

Formula: C₁₂H₁₅F₃N₂O
MDL Number: MFCD12832920
CAS Number: 1184392-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR