4-(4-Benzyl-1-piperazinyl)-3-(trifluoromethyl)phenylamine

051280-2.5G
101895-014EA 1783.78 USD
101895-014
4-(4-Benzyl-1-piperazinyl)-3-(trifluoromethyl)phenylamine
4-(4-Benzyl-1-piperazinyl)-3-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₈H₂₀F₃N₃
MDL Number: MFCD09909591
CAS Number: 946719-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR