N-1-Butyl-N-1-methyl-2-(trifluoromethyl)-1,4-benzenediamine

Supplier: Matrix Scientific

051274-2.5G 051274-500MG
101894-990EA 4660.85 USD
101894-990 101894-988
N-1-Butyl-N-1-methyl-2-(trifluoromethyl)-1,4-benzenediamine
N-1-Butyl-N-1-methyl-2-(trifluoromethyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₂H₁₇F₃N₂
MDL Number: MFCD12832921
CAS Number: 1184470-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR