2-[4-Amino(methyl)-2-(trifluoromethyl)anilino]-1-ethanol

Supplier: Matrix Scientific

051272-500MG 051272-2.5G
101894-980EA 538.46 USD
101894-980 101894-982
2-[4-Amino(methyl)-2-(trifluoromethyl)anilino]-1-ethanol
2-[4-Amino(methyl)-2-(trifluoromethyl)anilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₃F₃N₂O
MDL Number: MFCD12768354
CAS Number: 1183225-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR