N-1,N-1-Dimethyl-2-(trifluoromethyl)-1,4-benzenediamine

Supplier: Matrix Scientific

Synonyms: N1,N1-Dimethyl-2-(trifluoromethyl)benzene-1,4-diamine

051270-500MG 051270-2.5G
101894-972EA 364.62 USD
101894-972 101894-974
N-1,N-1-Dimethyl-2-(trifluoromethyl)-1,4-benzenediamine
N-1,N-1-Dimethyl-2-(trifluoromethyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₉H₁₁F₃N₂
MW: 204.2 g/mol
MDL Number: MFCD10686579
CAS Number: 54672-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR