4-[3,4-Dihydro-1(2H)-quinolinyl]aniline

Supplier: Matrix Scientific

051267-500MG 051267-2.5G
101894-960EA 364.62 USD
101894-960 101894-962
4-[3,4-Dihydro-1(2H)-quinolinyl]aniline
4-[3,4-Dihydro-1(2H)-quinolinyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₆N₂
MDL Number: MFCD08699238
CAS Number: 861038-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR