N-1-,N-1-Dipropyl-1,4-benzenediamine
Supplier: Matrix Scientific
Synonyms:
N1,N1-Dipropylbenzene-1,4-diamine
051259-2.5G
051259-500MG
101894-928EA
160
USD
101894-928
101894-930
N-1-,N-1-Dipropyl-1,4-benzenediamine
N-1-,N-1-Dipropyl-1,4-benzenediamine
Hazard: Irritant/Store in Freezer
Formula:
C₁₂H₂₀N₂ MW: 192.31 g/mol Boiling Pt: 155…156 °C (6 Torr) |
MDL Number:
MFCD09044836 CAS Number: 105293-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...