N-1-Butyl-N-1-methyl-1,4-benzenediamine

Supplier: Matrix Scientific

051253-500MG 051253-2.5G
101894-904EA 364.62 USD
101894-904 101894-906
N-1-Butyl-N-1-methyl-1,4-benzenediamine
N-1-Butyl-N-1-methyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₁H₁₈N₂
MDL Number: MFCD09803412
CAS Number: 104178-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR