N-1-Benzyl-2-chloro-N-1-ethyl-1,4-benzenediamine

Supplier: Matrix Scientific

051246-500MG 051246-2.5G
101894-876EA 364.62 USD
101894-876 101894-878
N-1-Benzyl-2-chloro-N-1-ethyl-1,4-benzenediamine
N-1-Benzyl-2-chloro-N-1-ethyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₅H₁₇ClN₂
MDL Number: MFCD07365140
CAS Number: 893750-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR