3-Chloro-4-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051232-500MG 051232-2.5G
101894-820EA 166.15 USD
101894-820 101894-822
3-Chloro-4-(2-methyl-1-piperidinyl)aniline
3-Chloro-4-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD07365134
CAS Number: 893750-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR