1-(4-Amino-2-chlorophenyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051228-500MG 051228-2.5G
101894-804EA 364.62 USD
101894-804 101894-806
1-(4-Amino-2-chlorophenyl)-3-pyrrolidinol
1-(4-Amino-2-chlorophenyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD13561459
CAS Number: 1220037-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR