2-(2-Amino-4-fluoromethylanilino)-1-ethanol

Supplier: Matrix Scientific

051201-2.5G 051201-500MG
101894-696EA 364.62 USD
101894-696 101894-698
2-(2-Amino-4-fluoromethylanilino)-1-ethanol
2-(2-Amino-4-fluoromethylanilino)-1-ethanol

Hazard: Irritant

Formula: C₉H₁₃FN₂O
MDL Number: MFCD11642964
CAS Number: 912284-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR