4-(2-Amino-4-methylphenyl)-2-piperazinone

Supplier: Matrix Scientific

051185-500MG 051185-2.5G
101894-632EA 364.62 USD
101894-632 101894-634
4-(2-Amino-4-methylphenyl)-2-piperazinone
4-(2-Amino-4-methylphenyl)-2-piperazinone

Hazard: Irritant

Formula: C₁₁H₁₅N₃O
MDL Number: MFCD11645432
CAS Number: 1097785-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR