2-(1-Azepanyl)-5-methylaniline

Supplier: Matrix Scientific

051184-500MG 051184-2.5G
101894-628EA 364.62 USD
101894-628 101894-630
2-(1-Azepanyl)-5-methylaniline
2-(1-Azepanyl)-5-methylaniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD10686555
CAS Number: 946731-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR