5-Methyl-2-(4-methyl-1-piperidinyl)aniline
Supplier: Matrix Scientific
051181-500MG
101894-616EA
461.54
USD
101894-616
5-Methyl-2-(4-methyl-1-piperidinyl)aniline
5-Methyl-2-(4-methyl-1-piperidinyl)aniline
Hazard: Irritant
Formula:
C₁₃H₂₀N₂ |
MDL Number:
MFCD11643966 CAS Number: 946719-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...