2-(2-Amino-4-dimethylanilino)-1-ethanol

Supplier: Matrix Scientific

051172-500MG 051172-2.5G
101894-580EA 364.62 USD
101894-580 101894-582
2-(2-Amino-4-dimethylanilino)-1-ethanol
2-(2-Amino-4-dimethylanilino)-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₆N₂O
MDL Number: MFCD11645572
CAS Number: 912284-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR