2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-(trifluoromethyl)aniline
Supplier: Matrix Scientific
051164-2.5G
051164-500MG
101894-550EA
838.46
USD
101894-550
101894-548
2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-(trifluoromethyl)aniline
2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-(trifluoromethyl)aniline
Hazard: Irritant
Formula:
C₁₆H₁₅F₃N₂ |
MDL Number:
MFCD00042163 CAS Number: 175134-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...