1-[2-Amino-4-(trifluoromethyl)phenyl]-4-piperidinol

Supplier: Matrix Scientific

051157-500MG 051157-2.5G
101894-520EA 364.62 USD
101894-520 101894-522
1-[2-Amino-4-(trifluoromethyl)phenyl]-4-piperidinol
1-[2-Amino-4-(trifluoromethyl)phenyl]-4-piperidinol

Hazard: Irritant

Formula: C₁₂H₁₅F₃N₂O
MDL Number: MFCD09738253
CAS Number: 954274-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR