5-Chloro-2-[3,4-dihydro-1(2H)-quinolinyl]aniline

Supplier: Matrix Scientific

051140-500MG 051140-2.5G
101894-452EA 364.62 USD
101894-452 101894-454
5-Chloro-2-[3,4-dihydro-1(2H)-quinolinyl]aniline
5-Chloro-2-[3,4-dihydro-1(2H)-quinolinyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂
MDL Number: MFCD08696897
CAS Number: 937606-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR