1-(2-Amino-4-chlorophenyl)-4-piperidinol
Supplier: Matrix Scientific
051131-2.5G
051131-500MG
101894-416EA
364.62
USD
101894-416
101894-418
1-(2-Amino-4-chlorophenyl)-4-piperidinol
1-(2-Amino-4-chlorophenyl)-4-piperidinol
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O |
MDL Number:
MFCD09731621 CAS Number: 953717-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...